Learn More
Grainger Metal Hose 6 ID x 10 ft.L
Supplier:  Grainger 16T871
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 6 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made from 304 stainless steel with a smooth bore. The hose has a temperature range of -40°F to 1000°F. It has male NPT threaded ends and is 30 feet in length. The hose is designed for conveying air, water, oil and other fluids in industrial applications. It is durable, flexible, and resistant to kinks. The interlocked construction allows it to handle high pressures.
Catalog No. 50-276-6135