Learn More
Grainger Metal Hose 6 ID x 14 ft.L
Supplier:  Grainger 16T820
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 6 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a corrugated interlocked design for flexibility. This hose has a temperature range of -40 to 450 degrees Fahrenheit. It has a zinc electroplated finish and is designed for use transferring hot air, fumes, and abrasive materials. The hose has a 6 inch NPT male end and 6 inch NPT swivel female end. It is 72 inches long with a bend radius of 12 inches. This durable metal hose is ideal for industrial applications requiring a flexible connection.
Catalog No. 50-276-6616