Learn More
Grainger Metal Hose 6 ID x 16 ft.L
Supplier:  Grainger 16T874
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 4 inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations which allow it to be very flexible. The hose is made from 304 stainless steel with a mirror finish. It has a 4 inch NPT male threaded end fitting on one side and a 4 inch NPT female threaded end fitting on the other side. The hose is 72 inches long when fully extended but can compress down to 24 inches. It weighs approximately 15 pounds. This high quality stainless steel metal hose is ideal for transferring air, water, oil and other fluids in industrial applications.
Catalog No. 50-276-6253