Learn More
Grainger Metal Hose 6 ID x 25 ft.L
Supplier:  Grainger 16T877
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 6 inch diameter interlocked metal hose with a working pressure of 300 psi. It has a stainless steel construction with a smooth bore interior and is designed for conveying air, water, oil and other fluids. This flexible metal hose can withstand temperatures from -40 to 300 degrees Fahrenheit. It has zinc-plated carbon steel fittings and is 25 feet in length. The interlocked annular corrugated stainless steel design allows flexibility while maintaining strength and durability. This industrial metal hose meets SAE 100R2 standards.
Catalog No. 50-276-5771