Learn More
Grainger Metal Hose 6 ID x 6 ft.L
Supplier:  Grainger 16T816
Encompass_Preferred
The Hose Master Flexible Metal Hose Assembly is a stainless steel braided hose with a 1/4" NPT male end and a 1/4" NPT female swivel end. It has a working pressure of up to 6000 psi and a temperature range of -40°F to 1000°F. The assembly is 36" long with a 1/4" diameter. It is made in the USA and is designed for use in hydraulic applications where flexibility and durability are needed. The stainless steel braiding provides strength while allowing for bending and vibration absorption. This high quality hose assembly meets SAE 100R2 standards.
Catalog No. 50-276-6749