Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-413
Used to identify property from components to finished products. Brady™ MetaLabel™ Series Metallic, Non-Metal Labels are made of Metallic, Non-Metal Polyester.
Supplier:  Brady™ THT184133
 View more versions of this product
Catalog No. 19-109-254
Add to Cart
Description
Description
  • Available in various sizes
  • Acrylic-based, industrial-strength adhesive for multiple applications
Specifications
Specifications
UL Recognised; cUL Recognised
Acrylic, Permanent
Light Gray
Roll
Single-side Printable
Label
Asset Tracking, General and Industrial Identificationentification, Rating Plates
1
THT-18-413-3
3000/Pk.
76.20 mm (W) x 25.40 mm (H)
-94°F to +230°F
0.0034 in.
28.58 mm
1 in.
76.2 mm
1 in.
THT-18-413-3
Thermal Transfer Printable Labels
None
Satin
Polyester (B-480)
Thermal Transfer
Non-sterile
BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
-70°C
3000 Label(s) / Roll
Rectangle
76.2 x 25.4 mm (W x H)
-70°C to +110°C
0.086 mm
25.40 x 76.20 mm
3 in.
76.20 mm
25.4 mm
Product Suggestions
Product Suggestions