Learn More
Medchemexpress LLC Metalaxyl-M | 70630-17-0 | 98.1% | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0843A10MM/1ML
Encompass_Preferred
Metalaxyl-M ((R)-Metalaxyl) is an orally active and selective inhibitor of fungal RNA polymerase. It exhibits fungicidal activity by selectively interfering with the synthesis of fungal ribosomal RNA. It can also be utilized to induce inflammation in hepatocytes, regulate tryptophan metabolism, and for ecotoxicology studies.
  • Orally active
  • Selective inhibitor of fungal RNA polymerase
  • Exhibits fungicidal activity
  • Interferes with fungal ribosomal RNA synthesis
  • Induces inflammation in hepatocytes
  • Regulates tryptophan metabolism
  • Used for ecotoxicology studies
Catalog No. 50-004-08161