Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metaldehyde | 108-62-3 | 98.0% | 500 MG
SDP

Supplier:  Medchemexpress LLC HYB1870500MG

Encompass_Preferred

Metaldehyde is a commonly used pesticide for mollusks such as slugs and snails.

  • Purity: 98.0%
  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 176.21
  • Formula: C8H16O4

Catalog No. 50-004-11049


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.