Learn More
Cayman Chemical MetalothIonIncod Polyclo 100uL
SDP
Supplier:  Cayman Chemical 172130100 UL
Encompass_Preferred
A selective inhibitor of TGF-B type 1 receptor (TGFBR1 or ALK5 IC50 18 nM) inhibits the expression of collagen type I in cells (IC50 93 nM) and in mice when given orally at 10 mg/kg once a day reduces typical features of fibrosis
Catalog No. 50-296-7918