Learn More
Medchemexpress LLC Metamizole sodium | 68-89-3 | MFCD00020783 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB1279A10MM/1ML
Encompass_Preferred
Metamizole sodium (Dipyrone) is an orally active cyclooxygenase (COX) inhibitor. It can inhibit cell proliferation and promote cell apoptosis. It also exhibits anti-inflammatory and antioxidant activities. Metamizole sodium is an antipyretic, analgesic, and spasmolytic agent, used in research to relieve various types of pain.
  • Orally active cyclooxygenase (COX) inhibitor
  • Inhibits cell proliferation and promotes cell apoptosis
  • Has anti-inflammatory and antioxidant activities
  • Antipyretic, analgesic and spasmolytic agent
  • Can be used in research to relieve a variety of pain
Catalog No. 50-004-13254