Learn More
Medchemexpress LLC Metamizole sodium (Dipyrone; Methamizole) | 68-89-3 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB1279A5G
Encompass_Preferred
Metamizole sodium is an orally active cyclooxygenase (COX) inhibitor. It can inhibit cell proliferation and promote cell apoptosis. It also possesses anti-inflammatory and antioxidant activities. Metamizole sodium is an antipyretic, analgesic, and spasmolytic agent, used in research to relieve various types of pain.
  • Orally active cyclooxygenase (COX) inhibitor.
  • Inhibits cell proliferation and promotes cell apoptosis.
  • Exhibits anti-inflammatory and antioxidant activities.
  • Functions as an antipyretic, analgesic, and spasmolytic agent.
  • Used in research for pain relief.
Catalog No. 50-004-12962