Learn More
Medchemexpress LLC Metamizole (sodium hydrate) | 5907-38-0 | 99.9% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB127910G
Encompass_Preferred
Metamizole sodium hydrate is an orally active cyclooxygenase (COX) inhibitor. It can inhibit cell proliferation and promote cell apoptosis. It also has anti-inflammatory and antioxidant activities, and acts as an antipyretic, analgesic, and spasmolytic agent. It can be used in research to relieve various types of pain.
  • Orally active cyclooxygenase (COX) inhibitor.
  • Inhibits cell proliferation and promotes cell apoptosis.
  • Possesses anti-inflammatory and antioxidant activities.
  • Acts as an antipyretic, analgesic, and spasmolytic agent.
  • Used in research to relieve various types of pain.
Catalog No. 50-004-12979