Learn More
Medchemexpress LLC Metamizole sodium hydrate | 5907-38-0 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB12795G
Encompass_Preferred
Metamizole sodium hydrate (Dipyrone) is an orally active cyclooxygenase (COX) inhibitor. It can inhibit cell proliferation and promote cell apoptosis, and possesses anti-inflammatory and antioxidant activities. It is also an antipyretic, analgesic, and spasmolytic agent, used in research to relieve various types of pain.
  • Orally active cyclooxygenase (COX) inhibitor
  • Inhibits cell proliferation and promotes cell apoptosis
  • Possesses anti-inflammatory and antioxidant activities
  • Antipyretic, analgesic, and spasmolytic agent
  • Used in research to alleviate various types of pain
Catalog No. 50-004-11594