Learn More
Cayman Chemical MetamIzolesodIum salt 25g
SDP
Supplier:  Cayman Chemical 1577125 G
Encompass_Preferred
A nonopioid prodrug; inhibits COX-1 and COX-2 activity (IC50 = ~150 UG/ml for both in cell-free assays), as well as reduces AITC-induced increases in intracellular calcium levels in isolated rat and mouse dorsal root ganglia and IMR-90 human fibroblasts (IC50s = 30-91 µM in patch-clamp assays); inhibits ADP- and epinephrine-induced platelet aggregation in hPRP; prevents fever induced by LPS, TNF-α, and IL-6, but not arachidonic acid or PGE2, in rats at 120 mg/kg; increases the paw withdrawal threshold in the paw pressure test in rat models of carrageenan-induced acute pain and postoperative incision pain at 250 and 500 mg/kg
Catalog No. 50-296-9764