Learn More
Cayman Chemical MetamIzolesodIum salt 50mg
SDP
Supplier:  Cayman Chemical 3299650 MG
Encompass_Preferred
An analytical reference standard categorized as an analgesic and anti-inflammatory agent; has been found as an adulterant in samples seized by law enforcement; intended for research and forensic applications
Catalog No. 50-295-0117