Learn More
Medchemexpress LLC Metanilic acid | 121-47-1 | MFCD00065345 | 98.0% | 173.19 g/mol | C6H7NO3S | 100 G

Supplier: Medchemexpress LLC HYW016836100G
Metanilic acid (3-aminobenzenesulfonic acid, CAS 121-47-1) is an aromatic sulfonic acid used as a research reagent for organic synthesis and bioactivity studies, including investigational anticancer research. It is a white solid with formula C6H7NO3S and molecular weight 173.19 g/mol, commonly supplied at ~98.0% purity for laboratory use.
- High purity (98.0%) for laboratory use.
- Aromatic sulfonic acid used in organic synthesis.
- Relevant to bioactivity and investigational anticancer studies.
- Provided with supporting documentation: datasheet, COA, and SDS.
- Available in common laboratory pack sizes such as 100 g and 500 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.