Learn More
Medchemexpress LLC Benzenemethanol, α-[(1S)-1-aminoethyl]-3-hydroxy-, (αR)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1) | 33402-03-8 | 99.9% | 100 MG
Supplier: Medchemexpress LLC HYB1116100MG
Metaraminol tartrate is an α-adrenergic agonist and a sympathomimetic amine that directly and indirectly affects adrenergic receptors, with alpha effects being predominant. It acts as a vasopressor agent and is for research use only.
- α-Adrenergic agonist
- Sympathomimetic amine
- Acts as a vasopressor agent
- High purity: 99.9%