Learn More
Medchemexpress LLC Metaraminol (tartrate) | 33402-03-8 | 99.9% | 1 ML
Supplier: Medchemexpress LLC HYB111610MM/1ML
Metaraminol tartrate is an α-adrenergic agonist and sympathomimetic amine that directly and indirectly affects adrenergic receptors. It acts as a vasopressor agent, primarily through its alpha effects, making it suitable for various research applications.
- Acts as an α-adrenergic agonist
- Functions as a sympathomimetic amine
- Directly and indirectly affects adrenergic receptors
- Exhibits predominant alpha effects
- Serves as a vasopressor agent
- High purity of 99.9%
- Solid appearance, white to off-white color