Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metaraminol tartrate | 33402-03-8 | 99.9% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYB111650MG

Encompass_Preferred

Metaraminol tartrate is an α-adrenergic agonist and a sympathomimetic amine that directly and indirectly affects adrenergic receptors with predominant alpha effects. It acts as a vasopressor agent and appears as a white to off-white solid.

  • α-adrenergic agonist
  • Sympathomimetic amine
  • Directly and indirectly affects adrenergic receptors with predominant alpha effects
  • Acts as a vasopressor agent
  • Molecular Weight: 317.29
  • Formula: C13H19NO8
  • Solubility in DMSO: 100 mg/mL (315.17 mM), may require ultrasonic
  • For research use only; not sold to patients
  • Controlled substance and may not be for sale in all territories

Catalog No. 50-004-11554


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.