Learn More
Medchemexpress LLC HY-120118 5mg Medchemexpress, Metarrestin CAS:1443414-10-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1201185MG
Encompass_Preferred
Medchemexpress, HY-120118 5mg Metarrestin CAS:1443414-10-5 Metarrestin (ML246) disrupts the nucleolar structure and inhibits RNA polymerase (Pol) I transcription, at least in part by interacting with the translation elongation factor eEF1A2. Metarrestin is a first-in-class perinucleolar compartment inhibitor with a favorable PK profile, which effectively suppresses metastasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9580