Learn More
Medchemexpress LLC Metergoline | 17692-51-2 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB10335MG
Encompass_Preferred
Metergoline is a serotonin (5-HT) receptor and dopamine receptors antagonist. Its pKis are 8.64, 8.75, and 8.75 for 5-HT2A, 5-HT2B, and 5-HT2C, respectively. It is also a high-affinity ligand for the h5-HT7 receptor, with a Ki of 16 nM. Additionally, Metergoline acts as a reversible neural Na+ channels inhibitor. It is commonly used in research for seasonal affective disorder and prolactin hormone regulation.
  • Serotonin (5-HT) receptor and dopamine receptor antagonist
  • High-affinity ligand for the h5-HT7 receptor (Ki = 16 nM)
  • Reversible neural Na+ channels inhibitor
  • Used in research for seasonal affective disorder and prolactin hormone regulation
Catalog No. 50-004-13801