Learn More
Medchemexpress LLC HY-110228 1mg Medchemexpress, Metformin D6 (hydrochloride) CAS:1185166-01-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1102281MG
Encompass_Preferred
Medchemexpress, HY-110228 1mg Metformin D6 (hydrochloride) CAS:1185166-01-1 Metformin D6 hydrochloride is a deuterium labeled Metformin hydrochloride. Metformin hydrochloride inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin hydrochloride triggers autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0781