Learn More
Cayman Chemical MetformIn-d6hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 169215 MG
Encompass_Preferred
An internal standard for the quantification of metformin by GC- or LC-MS
Catalog No. 50-296-5220