Learn More
Medchemexpress LLC Methacrylic anhydride | 760-93-0 | MFCD00008586 | 98.8% | 154.16 g/mol | C8H10O3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0173305G
Encompass_Preferred
Methacrylic anhydride is a reactive anhydride (2-propenoic acid, 2-methyl-, 1,1'-anhydride) used as a polymerizable reagent and acylating agent in organic synthesis and polymer chemistry. It is commonly used to introduce methacryloyl functionality for polymer modification, biomaterial functionalization, and crosslinking reactions. Consult the batch COA for batch-specific purity and handling instructions.
  • Reactive anhydride for introducing methacryloyl groups.
  • Suitable for polymer synthesis, modification, and crosslinking.
  • High purity, with batch-specific purity reported on COA.
  • Compatible with common organic solvents and acylation protocols.
  • Requires careful handling due to reactivity and moisture sensitivity.
  • Available in multiple package sizes for research use.
Catalog No. 50-003-01867