Learn More
Medchemexpress LLC Methacrylic anhydride | 760-93-0 | MFCD00008586 | 98.8% | 154.17 g/mol | C8H10O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01733010MM/1ML
Encompass_Preferred
Methacrylic anhydride is a reactive methacrylation reagent used to introduce methacrylate groups onto nucleophiles such as amines and hydroxyls. Supplied as a 10 mM solution in DMSO (1 mL), it is used for polymer modification, biomaterial functionalization, and small-molecule synthesis; handle and store cold to minimize spontaneous polymerization.
  • Reactive anhydride for methacrylation of amines and alcohols.
  • Supplied as a ready-to-use 10 mM solution in DMSO for quick setup.
  • High purity reduces side reactions and improves reproducibility.
  • Suitable for polymer modification, biomaterial functionalization, and synthesis.
  • Provided with COA and SDS for quality and safety traceability.
  • Recommended cold storage to limit spontaneous polymerization.
Catalog No. 50-003-01187