Learn More
Medchemexpress LLC Methanamine, N-(2,3-diphenyl-1,2,4-thiadiazol-5(2H)-ylidene)-, hydrobromide | 265980-25-4 | 96.81% | 348.26 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY11001210MG
Encompass_Preferred
SCH-202676 hydrobromide is an allosteric modulator of G protein-coupled receptors (GPCRs) and adenosine receptor (AR). It has antiviral activity and inhibits 3CLpro in a time-dependent manner with an IC50 value of 0.655 μM.
  • Allosteric modulator of G protein-coupled receptors (GPCRs) and adenosine receptor (AR).
  • Has antiviral activity.
  • Inhibits 3CLpro in a time-dependent manner with an IC50 value of 0.655 μM.
Catalog No. 50-003-44719