Learn More
Medchemexpress LLC Methanaminium, 1-carboxy-N,N,N-trimethyl-, chloride (1:1) | 590-46-5 | 153.61 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN073910MM/1ML
Encompass_Preferred
Betaine chloride is an analytical standard intended for research and analytical applications. This natural compound, also known as betaine hydrochloride, functions as an active methyl-donor that helps maintain normal DNA methylation patterns.
  • Analytical standard for assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-004-16779