Learn More
Supply Solutions Burdick & Jackson - Methanol, BioSyn. Package Size: 56L. Container Type:SSPDS. Suggested Applications: Biosynthesis, Applications requiring low water content
Supplier:  Supply Solutions BJ BC23356
Encompass_Preferred
67-56-1, Methanol, BioSynr. Suggested Application: Biosynthesis, Applications requiring low water content. Package Size: 56L. Container type is SSPDS. Hazard Class: 3. Package Group: II. Molecular Weight: 32.04. Density (g/mL) at 20 deg C: 0.7913. Boiling point (deg C): 64.7. Freezing point (deg C): -97.7. Flashpoint (deg C): 11. Manufacturer Name: Burdick and Jackson. Manufacturer Part Number: BC233-56.; Size: 56L
Catalog No. 50-877-249