Learn More
Aqua Solutions METHANOL CARBONYL FREE 1LTR
SDP
Supplier:  Aqua Solutions 56001L
Encompass_Preferred
Methanol(Methyl Alcohol) Carbonyl Free, Formula: CH OH, Components: Methanol(Methyl Alcohol)(Low Acetone) CAS No.: 67-56-1, 100%, Health: 3, Flammability: 3, Reactivity: 0, Melting Point: -144 deg.F, Specific Gravity: 0.792
Catalog No. NC9198701
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov