Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol d4 99.8 atom D c50G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 34380350G
Encompass
Methanol-d4 is a deuterated NMR solvent containing 0.03 % (v/v) TMS (trimethylsilane). It is useful in NMR-based research and analyses.
Catalog No. 50-290-2898