Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol d4 99 atom D
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 42287850ML
Encompass
Methanol-d4 (CD3OD) is a deuterated NMR solvent. It is useful in NMR-based research and analyses. Absolute infrared absorption intensities of CD3OD have been reported between 8000 and 350cm-1.1
Catalog No. 50-290-3506