Learn More
Cambridge Isotope Laboratories Methanol-D4 (d, 99.8%) +0.05% V/v Tms, 811-98-3, MFCD00044637, 10x0.6 mL
Supplier:  Cambridge Isotope Laboratories DLM24TB10X0.6
Encompass_Preferred
Chemical Formula: CD3OD, MW: 36.07, 99.8%.
Catalog No. NC9153994