Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol-OD 99.5 atom D100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 151939100G
Encompass
Methan(ol-d) (Methanol-OD CH3OD) is a deuterated form of methanol. Dissociation of CH3OD adsorbed on the clean W(100) surface resulted in the formation of hydrogen carbon and oxygen (-CO). Additionally carbon dioxide water and methyl formate were obtained when similar dissociation was conducted over the W(100)-(5x1)C surface. The dissociation process was monitored by temperature-programmed reaction spectroscopy. Self-diffusion coefficient shear viscosities and densities of liquid CH3OD have been determined by NMR spin-echo technique.
Catalog No. 50-289-9402