Learn More
Medchemexpress LLC Methanone, 2-benzofuranyl(4-nitrophenyl)- | 93944-58-2 | 99.49% | 267.24 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1490085MG
Encompass_Preferred
α-Amylase-IN-3 (Compound 4) is an inhibitor for α-Amylase, AChE, and BChE, with IC50s of 18.04 μM, 21.04 μM, and 22.2 μM, respectively. It exhibits antioxidant activity and can be used in research related to diabetes and oxidative stress-associated diseases. This product is provided as a white to off-white solid.
  • Inhibitor for α-Amylase, AChE, and BChE
  • Exhibits antioxidant activity
  • Used in research for diabetes
  • Used in research for oxidative stress-associated diseases
  • White to off-white solid appearance
Catalog No. 50-003-60900