Learn More
Medchemexpress LLC Methanone, [6-methoxy-2-[4-methoxy-3-(phosphonooxy)phenyl]-1H-indol-3-yl](3,4,5-trimethoxyphenyl) | 288847-41-6 | 99.5% | 587.42 g·mol⁻1 | C26H24NNa2O10P | 100MG

Supplier: Medchemexpress LLC HY123283100MG
OXi8007 is a water-soluble phosphate prodrug that acts as a tubulin/microtubule-binding inhibitor for research applications. It is supplied as the disodium phosphate salt with high analytical purity and is intended for in vitro research use only. Key chemical identifiers include CAS 288847-41-6, formula C26H24NNa2O10P, and molecular weight 587.42 g·mol⁻1.
- Tubulin/microtubule binding mechanism suitable for anti-microtubule studies.
- High purity: 99.5%.
- Solid, off-white to light yellow appearance; phosphate salt increases aqueous solubility.
- Chemical formula C26H24NNa2O10P and molecular weight 587.42 g·mol⁻1.
- Available in multiple solid masses and solution formats for assay flexibility.
- Recommended storage: 4°C sealed; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month), sealed, away from moisture and light.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.