Learn More
Medchemexpress LLC Methanone, bis(2,4-dihydroxyphenyl)- | 131-55-5 | MFCD08692012 | 99.7% | 246.22 g/mol | C13H10O5 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW03612050G
Encompass_Preferred
Benzophenone-2 (2,2',4,4'-tetrahydroxybenzophenone) is a research-grade UV-filter compound used to study nuclear receptor activation and CYP3A regulation. It modulates the pregnane X receptor (PXR) and estrogen receptor α (ERα) and is intended for laboratory research use only, not for clinical or human use.
  • Acts as an endocrine disruptor targeting PXR and ERα.
  • High purity suitable for analytical and research applications.
  • Molecular formula C13H10O5; molecular weight 246.22 g/mol.
  • Available in multiple pack sizes to support varied experimental needs.
  • Supplied with batch-specific certificate of analysis for quality verification.
Catalog No. 50-003-01550