Learn More
Cayman Chemical Methedrone hydrochloride
SDP
Supplier:  Cayman Chemical 105295 MG
Encompass_Preferred
A phenethylamine, amphetamine, and cathinone derivative that acts as a triple reuptake/release/reversible MAO inhibitor; intended for use as a standard for the forensic analysis of samples that may contain this compound
Catalog No. 50-288-7865