Learn More
Cayman Chemical MethIcIlInsodIum salt 50mg
SDP
Supplier:  Cayman Chemical 2100750 MG
Encompass_Preferred
A semisynthetic penicillin antibiotic that can inhibit bacterial cell wall synthesis; used to study methicillin-resistance in S. aureus
Catalog No. 50-298-4333