Learn More
Sigma Aldrich Fine Chemicals Biosciences Methional 97 fG1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W2747041KGK
Encompass
Methional is the main sulfur-containing flavor compound in potatoes.
Catalog No. 50-290-2333