Learn More
Medchemexpress LLC Methionine | 00-00-0 | MFCD11983488 | 98.0% | C5H7D4NO2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13694S110MG
Encompass_Preferred
Methionine-d4 is the deuterium-labeled form of methionine used as an isotopic tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. The labeled compound provides accurate mass discrimination and consistent performance in analytical assays, and is supplied as a high-purity solid suitable for storage and routine laboratory use.
  • Deuterium-labeled internal standard for quantitative methods
  • Compatible with NMR, GC-MS, and LC-MS analyses
  • High chemical purity for reliable quantitation
  • Stable solid form for long-term storage
  • Suitable as a tracer in metabolic and pharmacokinetic studies
Catalog No. 50-002-59925