Learn More
Medchemexpress LLC Methionine-d3 | 284665-18-5 | 99.2% | 152.23 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13694S
Encompass_Preferred
Methionine-d3 is the deuterium-labeled Methionine. Methionine is an effective chemoprotective agent that can also inhibit neuronal activity through GABAA receptor activation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-52339