Learn More
Medchemexpress LLC Methionine-d3 | 284665-18-5 | MFCD00211078 | 99.2% | 152.23 g/mol | C5H8D3NO2S | 5 MG

Supplier: Medchemexpress LLC HY13694S5MG
Methionine-d3 is the deuterium-labeled analogue of methionine used as a stable isotope internal standard for mass spectrometry, tracer studies, and quantitative analytical assays. Supplied as a solid with high isotopic and chemical purity, it enables accurate quantitation and metabolic tracing in biochemical and pharmacokinetic workflows.
- Deuterium-labeled internal standard for mass spectrometry and tracer experiments.
- High chemical purity (99.2%).
- Molecular formula C5H8D3NO2S; molecular weight 152.23 g/mol.
- Available in small research pack sizes suitable for analytical work.
- Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.