Learn More
Medchemexpress LLC Methionine-d3 | 284665-18-5 | MFCD00211078 | 99.2% | C5H8D3NO2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13694S10MG
Encompass_Preferred
Methionine-d3 is a deuterium-labeled form of the amino acid methionine provided as a white to off-white solid. It serves as a stable-isotope internal standard for mass spectrometry, tracer studies, and analytical workflows requiring an isotopically labeled amino acid. The material offers high purity and defined storage stability, and is supplied in small milligram quantities for laboratory use.
  • High purity (99.2%).
  • Deuterium-labeled methyl group for mass-shifted internal standardization.
  • Suitable for mass spectrometry and tracer studies.
  • White to off-white solid form for easy handling and weighing.
  • Stable when stored under recommended conditions for reliable performance.
Catalog No. 50-002-59021