Learn More
American Radiolabeled Chemicals Inc METHIONINE L-ME-3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 0169250UC
Encompass_Preferred
Methonine, L-methyl-3H 250uCi in ethanol:water (7:3), 1 mCi/ml, S.A. 30-80 Ci/mmol, M.W. 149.2, Shipped in Dry Ice
Catalog No. 50-901-04311