Learn More
Medchemexpress LLC Methionine, N-[(1,1-dimethylethoxy)carbonyl]- | 93000-03-4 | MFCD00190811 | 95.0% | 249.33 | C10H19NO4S | 25 G

Supplier: Medchemexpress LLC 25GHYW052309
(tert-Butoxycarbonyl)methionine (Boc-methionine) is an N-protected methionine derivative used as a protected amino acid building block in peptide synthesis and related organic synthesis. It is supplied as a solid, white to off-white powder and is used when temporary protection of the amino group is required.
- N-Boc protected methionine suitable for peptide coupling and synthesis applications.
- Supplied as a solid, white to off-white powder.
- Purity 95.0%.
- Molecular weight 249.33 g/mol.
- Chemical formula C10H19NO4S.
- Storage: powder at -20°C (3 years) or 4°C (2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.