Learn More
Medchemexpress LLC Methionine sulfoxide | 62697-73-8 | 99.8% | 165.21 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW010104A1G
Encompass_Preferred
Methionine sulfoxide is an oxidation product of methionine with reactive oxygen species and can be regarded as a biomarker of oxidative stress in vivo.
  • Endogenous metabolite
  • Solid appearance
  • White to off-white color
  • Initial source: microorganisms
  • Structure classification: ketones, aldehydes, acids
Catalog No. 50-004-51692