Learn More
Medchemexpress LLC Methocarbamol | 532-03-6 | >99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB026250MG
Encompass_Preferred
Methocarbamol is an orally active central muscle relaxant. It blocks the muscular Nav1.4 channel and reversibly affects its voltage dependence of inactivation.
  • Potential for research into muscle spasms and pain syndromes
  • Orally active central muscle relaxant
  • Blocks muscular Nav1.4 channel
Catalog No. 50-004-05660