Learn More
Chempoint Com METHOCEL E4M Premium, Hydroxypropyl Methylcellulose, 50 lbs, Water-soluble, Viscosity 4000 cPs (typical)
Supplier:  Chempoint Com 0130564
Encompass
METHOCEL E4M Premium is a high-quality, water-soluble hydroxypropyl methylcellulose (HPMC) cellulose ether polymer suitable for various scientific and industrial applications. It is known for its exceptional rheology modification, thickening, and film-forming capabilities. This non-ionic polymer provides high compatibility with organic solvents and excellent pH stability. Its unique properties include reversible thermal gelation above 64°C, making it ideal for enhancing adhesion and improving formulation strength.
  • High-quality hydroxypropyl methylcellulose polymer
  • Compatible with some organic solvents
  • Excellent pH stability range of 2-13
  • Reversible thermal gelation above 64°C
  • Efficient at low concentrations for thickening
  • Used in pharmaceuticals and coatings
  • Kosher certified for Passover and Pareve
Catalog No. NC2059816