Learn More
Medchemexpress LLC Methoctramine tetrah 5mg | 104807-46-7 | 728.75 g/mol | C36H66Cl4N4O2 | 5 MG

Supplier: Medchemexpress LLC HY116294A5MG
Methoctramine tetrahydrochloride is the tetrahydrochloride salt of methoctramine, a selective M2 muscarinic receptor antagonist used in pharmacological research to probe M2 receptor-mediated signaling and cardiovascular responses. The compound is provided as a solid with high purity and documented storage and solubility properties.
- Selective M2 muscarinic receptor antagonist.
- Intended for research use in pharmacology and cardiovascular studies.
- Tetrahydrochloride salt; white to off-white solid.
- Purity: 97%.
- Molecular weight: 728.75 g/mol; formula: C36H66Cl4N4O2.
- Solubility: water 20 mg/mL (may require ultrasonic).
- Storage: -20°C sealed; in solution -80°C (6 months), -20°C (1 month).
- Typical laboratory pack size: 5 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.