Learn More
STEMCELL Technologies MethoCult™ SF H4436, Size: 100 mL
SDP
Supplier:  STEMCELL Technologies 04436
Encompass
MethoCult™ SF H4436 is a complete serum-free methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. MethoCult™ SF H4436 is formulated to support optimal growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage, megakaryocyte progenitors (CFU-GEMM) under defined serum-free conditions.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay and explore its utility as part of the cell therapy workflow (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/cfu-cell-therapy-questions.html#section-applications).
Catalog No. 50-197-4315